AMBH324A5609 Ambeed, Inc.

Irinotecan hydrochloride

  • MDL number MFCD01862255
  • Cas Number : 100286-90-6
  • Molecular Weight : 623.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/100286-90-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/100286-90-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart