AMBH324A560F Ambeed, Inc.

Dideoxycytidine

  • MDL number MFCD00012188
  • Cas Number : 7481-89-2
  • Molecular Weight : 211.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/7481-89-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/7481-89-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart