AMBH324A5618 Ambeed, Inc.

(1R,2S)-2-(3,4-Difluorophenyl)cyclopropanamine (R)-2-hydroxy-2-phenylacetate

  • MDL number MFCD22200985
  • Cas Number : 376608-71-8
  • Molecular Weight : 321.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/376608-71-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/376608-71-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart