AMBH324A561C Ambeed, Inc.

(2S,4R)-tert-Butyl 4-hydroxy-2-methylpyrrolidine-1-carboxylate

  • MDL number MFCD11111254
  • Cas Number : 114676-61-8
  • Molecular Weight : 201.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/114676-61-8.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/114676-61-8.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart