AMBH324A564F Ambeed, Inc.

(3R,4R)-tert-Butyl 3-amino-4-methoxypyrrolidine-1-carboxylate

  • MDL number MFCD20230534
  • Cas Number : 1400562-12-0
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1400562-12-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1400562-12-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart