AMBH324A5654 Ambeed, Inc.

(3R,7AR)-3-(tert-butyl)-7a-vinyldihydropyrrolo[1,2-c]oxazole-1,5(3H,6H)-dione

  • MDL number MFCD20040514
  • Cas Number : 1214741-20-4
  • Molecular Weight : 223.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1214741-20-4.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/1214741-20-4.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart