AMBH324A5669 Ambeed, Inc.

(R)-2-Benzyl-4-(tert-butoxy)-4-oxobutanoic acid

  • MDL number MFCD12755810
  • Cas Number : 122225-33-6
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/122225-33-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/122225-33-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart