AMBH324A5683 Ambeed, Inc.

Maraviroc

  • MDL number MFCD13188530
  • Cas Number : 376348-65-1
  • Molecular Weight : 513.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/376348-65-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/376348-65-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart