AMBH324A5685 Ambeed, Inc.

(E)-Ethyl 3-acetamidobut-2-enoate

  • MDL number MFCD02974463
  • Cas Number : 23652-67-7
  • Molecular Weight : 171.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/23652-67-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/23652-67-7.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart