AMBH324A5692 Ambeed, Inc.

(4S,5R)-3-Benzoyl-2,2-dimethyl-4-phenyloxazolidine-5-carboxylic acid

  • MDL number MFCD11046552
  • Cas Number : 153652-70-1
  • Molecular Weight : 325.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/153652-70-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/153652-70-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart