AMBH324A5695 Ambeed, Inc.

(E)-1-Bromo-4-(2-nitroprop-1-en-1-yl)benzene

  • MDL number MFCD01317591
  • Cas Number : 131981-75-4
  • Molecular Weight : 242.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/131981-75-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/131981-75-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart