AMBH324A56A0 Ambeed, Inc.

(2R,4R)-1-Boc-2-Hydroxymethyl-4-aminopyrrolidine hydrochloride

  • MDL number MFCD11101163
  • Cas Number : 1161931-71-0
  • Molecular Weight : 252.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1161931-71-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1161931-71-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart