AMBH324A56A1 Ambeed, Inc.

(2R,3R,4S,5R)-6-Oxohexane-1,2,3,4,5-pentayl pentaacetate

  • MDL number MFCD09996912
  • Cas Number : 3891-59-6
  • Molecular Weight : 390.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/3891-59-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/3891-59-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart