AMBH324A56A2 Ambeed, Inc.

Boc-D-Ser(tBu)-OH.DCHA

  • MDL number MFCD00236872
  • Cas Number : 248921-67-7
  • Molecular Weight : 442.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/248921-67-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/248921-67-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart