AMBH324A56A3 Ambeed, Inc.

(Z)-3-(1H-Indol-3-yl)acrylonitrile

  • MDL number MFCD28125617
  • Cas Number : 85452-79-5
  • Molecular Weight : 168.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/85452-79-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/85452-79-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart