AMBH324A56A5 Ambeed, Inc.

3-Cl-Tyr-OH

  • MDL number MFCD00045865
  • Cas Number : 7423-93-0
  • Molecular Weight : 215.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/7423-93-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/7423-93-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart