AMBH324A56B3 Ambeed, Inc.

Boc-Dab(Z)-OH.DCHA

  • MDL number MFCD00236879
  • Cas Number : 16947-89-0
  • Molecular Weight : 533.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/16947-89-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/16947-89-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart