AMBH324A56B9 Ambeed, Inc.

4-((1r,4r)-4-Ethylcyclohexyl)benzoic acid

  • MDL number MFCD06658176
  • Cas Number : 87592-41-4
  • Molecular Weight : 232.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/87592-41-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/87592-41-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart