AMBH324A56BA Ambeed, Inc.

7-Amino-3-Cephem-4-Carboxylic Acid

  • MDL number MFCD08063340
  • Cas Number : 36923-17-8
  • Molecular Weight : 200.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/36923-17-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/36923-17-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart