AMBH324A56BB Ambeed, Inc.

(S)-(4-(2-((tert-Butoxycarbonyl)amino)-3-methoxy-3-oxopropyl)phenyl)boronic acid

  • MDL number MFCD10698492
  • Cas Number : 224824-22-0
  • Molecular Weight : 323.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/224824-22-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/224824-22-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart