AMBH324A56BD Ambeed, Inc.

(R)-6-(3-Methylpiperazin-1-yl)nicotinonitrile

  • MDL number MFCD17488802
  • Cas Number : 1057682-05-9
  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1057682-05-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/1057682-05-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart