AMBH324A56C9 Ambeed, Inc.

Z-Pyr-OtBu

  • MDL number MFCD11975830
  • Cas Number : 81470-51-1
  • Molecular Weight : 319.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/81470-51-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/81470-51-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart