AMBH324A56D0 Ambeed, Inc.

3,4,5-Trifluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

  • MDL number MFCD09839001
  • Cas Number : 132123-39-8
  • Molecular Weight : 332.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/132123-39-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/132123-39-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart