AMBH324A56DD Ambeed, Inc.

(R)-2-(2-Hydroxy-3-((4-(3-oxomorpholino)phenyl)amino)propyl)isoindoline-1,3-dione

  • MDL number MFCD11977664
  • Cas Number : 446292-07-5
  • Molecular Weight : 395.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/446292-07-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/446292-07-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart