AMBH324A56E6 Ambeed, Inc.

(3R,4R)-1-Benzylpyrrolidine-3,4-diol

  • MDL number MFCD01073894
  • Cas Number : 163439-82-5
  • Molecular Weight : 193.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/163439-82-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/163439-82-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart