AMBH324A56EF Ambeed, Inc.

(4-(trans-4-Pentylcyclohexyl)phenyl)boronic acid

  • MDL number MFCD09750938
  • Cas Number : 143651-26-7
  • Molecular Weight : 274.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/143651-26-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/143651-26-7.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart