AMBH324A56F9 Ambeed, Inc.

Boc-L-Serine-beta-Lactone

  • MDL number MFCD01318414
  • Cas Number : 98541-64-1
  • Molecular Weight : 187.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/98541-64-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/98541-64-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart