AMBH324A56FC Ambeed, Inc.

4-(N-((4-Amino-2-methylpyrimidin-5-yl)methyl)formamido)-3-(benzoylthio)pent-3-en-1-yl benzoate

  • MDL number MFCD00867787
  • Cas Number : 299-88-7
  • Molecular Weight : 490.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/299-88-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/299-88-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart