AMBH324A5702 Ambeed, Inc.

Fmoc-Phe(4-NH2)-OH

  • MDL number MFCD00237664
  • Cas Number : 95753-56-3
  • Molecular Weight : 402.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/95753-56-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/95753-56-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart