AMBH324A5707 Ambeed, Inc.

trans,trans-4'-Propyl-4-(3,4,5-trifluorophenyl)bicyclohexyl

  • MDL number MFCD09838998
  • Cas Number : 131819-23-3
  • Molecular Weight : 338.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/131819-23-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/131819-23-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart