AMBH324A570B Ambeed, Inc.

(R)-tert-Butyl 1-(benzylamino)-3-methoxy-1-oxopropan-2-ylcarbamate

  • MDL number MFCD16620517
  • Cas Number : 880468-89-3
  • Molecular Weight : 308.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/880468-89-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/880468-89-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart