AMBH324A5714 Ambeed, Inc.

(R)-4-Amino-5-methoxy-5-oxopentanoic acid hydrochloride

  • MDL number MFCD28041186
  • Cas Number : 187458-77-1
  • Molecular Weight : 197.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/187458-77-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/187458-77-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart