AMBH324A5717 Ambeed, Inc.

(S)-Ethyl 4-amino-2-((tert-butoxycarbonyl)amino)-4-oxobutanoate

  • MDL number MFCD15141969
  • Cas Number : 84787-81-5
  • Molecular Weight : 260.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/84787-81-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/84787-81-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart