AMBH324A5730 Ambeed, Inc.

Chlorprothixene

  • MDL number MFCD00869180
  • Cas Number : 113-59-7
  • Molecular Weight : 315.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/113-59-7.html

Safety Information

signalword Danger
pcodes P301+P310
MSDS https://www.ambeed.com/products/113-59-7.html
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart