AMBH324A573C Ambeed, Inc.

Ac-Glu-Glu-Met-Gln-Arg-Arg-Ala-Asp-NH2

  • MDL number MFCD18072161
  • Cas Number : 868844-74-0
  • Molecular Weight : 1075.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/868844-74-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/868844-74-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart