AMBH324A5741 Ambeed, Inc.

(R)-2-((tert-Butoxycarbonyl)amino)-3-hydroxy-3-methylbutanoic acid

  • MDL number MFCD02682576
  • Cas Number : 288159-40-0
  • Molecular Weight : 233.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/288159-40-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/288159-40-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart