AMBH324A574B Ambeed, Inc.

Adenosine, N-benzoyl-5'-O-[bis(4-methoxyphenyl)phenylmethyl]-3'-O-[(1,1-dimethylethyl)dimethylsilyl]-, 2'-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

  • MDL number MFCD00274109
  • Cas Number : 129451-75-8
  • Molecular Weight : 988.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/129451-75-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/129451-75-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart