AMBH324A574C Ambeed, Inc.

(1R,2R)-N1,N1,N2,N2-Tetramethylcyclohexane-1,2-diamine

  • MDL number MFCD03411208
  • Cas Number : 53152-69-5
  • Molecular Weight : 170.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/53152-69-5.html

Safety Information

signalword Warning
pcodes P261-P301+P310-P305+P351+P338
MSDS https://www.ambeed.com/products/53152-69-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart