AMBH324A574D Ambeed, Inc.

BIBR 1532

  • MDL number MFCD11112195
  • Cas Number : 321674-73-1
  • Molecular Weight : 331.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/321674-73-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/321674-73-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart