AMBH324A574F Ambeed, Inc.

Dichloro(1,5-cyclooctadiene)platinum(II)

  • MDL number MFCD00012413
  • Cas Number : 12080-32-9
  • Molecular Weight : 374.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/12080-32-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/12080-32-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart