AMBH324A5761 Ambeed, Inc.

Dinoprost tromethamine

  • MDL number MFCD00077863
  • Cas Number : 38562-01-5
  • Molecular Weight : 475.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/38562-01-5.html

Safety Information

signalword Danger
pcodes P201-P280-P301+P312+P330-P308+P313
MSDS https://www.ambeed.com/products/38562-01-5.html
pictograms GHS07,GHS08
hstatements H302-H360
Availability details for
Quantity
  • Availability
  • Add to Cart