AMBH324A5771 Ambeed, Inc.

(1R,3S)-rel-Cyclohexane-1,3-diamine dihydrochloride

  • MDL number MFCD26406404
  • Cas Number : 498532-32-4
  • Molecular Weight : 187.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/498532-32-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/498532-32-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart