AMBH324A5772 Ambeed, Inc.

(S)-2-((tert-Butoxycarbonyl)amino)-4-(tritylthio)butanoic acid

  • MDL number MFCD00672309
  • Cas Number : 201419-16-1
  • Molecular Weight : 477.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/201419-16-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/201419-16-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart