AMBH324A577D Ambeed, Inc.

N-Acetyl-D-glucosamine

  • MDL number MFCD00136044
  • Cas Number : 7512-17-6
  • Molecular Weight : 221.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/7512-17-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/7512-17-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart