AMBH324A577E Ambeed, Inc.

(S)-2-((Methoxycarbonyl)amino)-3,3-dimethylbutanoic acid

  • MDL number MFCD07778455
  • Cas Number : 162537-11-3
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/162537-11-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/162537-11-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart