AMBH324A5780 Ambeed, Inc.

(S)-tert-Butyl 2-((tert-butoxycarbonyl)amino)-4-hydroxybutanoate

  • MDL number MFCD08275190
  • Cas Number : 81323-58-2
  • Molecular Weight : 275.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/81323-58-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/81323-58-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart