AMBH324A578A Ambeed, Inc.

4-(4-(Dimethylamino)phenyl)but-3-en-2-one

  • MDL number MFCD00052839
  • Cas Number : 5432-53-1
  • Molecular Weight : 189.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/5432-53-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/5432-53-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart