AMBH324A578B Ambeed, Inc.

Adenosine 5'-diphosphate bis(cyclohexylammonium) salt

  • MDL number MFCD00058338
  • Cas Number : 102029-87-8
  • Molecular Weight : 625.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/102029-87-8.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/102029-87-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart