AMBH324A579E Ambeed, Inc.

(2S,4S)-tert-Butyl 2-cyano-4-fluoropyrrolidine-1-carboxylate

  • MDL number MFCD09263600
  • Cas Number : 426844-76-0
  • Molecular Weight : 214.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/426844-76-0.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/426844-76-0.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart