AMBH324A579F Ambeed, Inc.

Z-Arg-OH

  • MDL number MFCD00001762
  • Cas Number : 1234-35-1
  • Molecular Weight : 308.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1234-35-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1234-35-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart